Product Details

SNP ID
rs61740769
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:115962720 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGCTGGCGCTGGCCGTACTCGCC[G/T]CCTTGTACGGCCACTGCGAGCGCGT
Phenotype
MIM: 610046
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LVRN PubMed Links

Gene Details

Gene
LVRN
Gene Name
laeverin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173800.4 281 Missense Mutation GCC,TCC A35S NP_776161.3
XM_011543263.2 281 Missense Mutation GCC,TCC A35S XP_011541565.1
XM_011543264.2 281 Missense Mutation GCC,TCC A35S XP_011541566.1
XM_017009215.1 281 Missense Mutation GCC,TCC A35S XP_016864704.1

View Full Product Details