Product Details

SNP ID
rs61749473
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:107504819 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGGGTCCTGGGATCCTATGGGAC[A/C]AAATGGCTCAACAGGTTTGCCTTTA
Phenotype
MIM: 613306
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ALKBH8 PubMed Links

Gene Details

Gene
ALKBH8
Gene Name
alkB homolog 8, tRNA methyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301010.1 1749 Missense Mutation GGT,TGT G615C NP_001287939.1
NM_138775.2 1749 Missense Mutation GGT,TGT G612C NP_620130.2
XM_005271728.2 1749 Missense Mutation GGT,TGT G475C XP_005271785.1
XM_005271729.3 1749 Missense Mutation GGT,TGT G475C XP_005271786.1
XM_017018553.1 1749 Missense Mutation GGT,TGT G565C XP_016874042.1
XM_017018554.1 1749 Missense Mutation GGT,TGT G562C XP_016874043.1
XM_017018555.1 1749 Missense Mutation GGT,TGT G360C XP_016874044.1
XM_017018556.1 1749 Missense Mutation GGT,TGT G310C XP_016874045.1
XM_017018557.1 1749 Intron XP_016874046.1

View Full Product Details