Product Details

SNP ID
rs58560242
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:80410359 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTGCTAGTGCAGGAGGCAGTGTT[A/G]GCTGCTACAGGAATAGCATGGAGCG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM213A PubMed Links

Gene Details

Gene
FAM213A
Gene Name
family with sequence similarity 213 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243778.1 Intron NP_001230707.1
NM_001243779.1 Intron NP_001230708.1
NM_001243780.1 Intron NP_001230709.1
NM_001243781.1 Intron NP_001230710.1
NM_001243782.1 Intron NP_001230711.1
NM_032333.4 Intron NP_115709.3
XM_011540265.2 Intron XP_011538567.1
XM_011540266.2 Intron XP_011538568.1
XM_017016775.1 Intron XP_016872264.1

View Full Product Details