Product Details

SNP ID
rs73661313
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:6950731 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTTCTCTCTTCCGGAAGCTCAGC[G/T]GAAGCAGTATGGGTAGCGTACTCCA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DEFA8P PubMed Links

Gene Details

Gene
DEFA8P
Gene Name
defensin alpha 8, pseudogene
There are no transcripts associated with this gene.

Gene
DEFA9P
Gene Name
defensin alpha 9, pseudogene
There are no transcripts associated with this gene.

View Full Product Details