Product Details

SNP ID
rs12675997
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:56301871 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACAGAGGAAGCAGTGAAGTCTTGTC[C/G]TGTAGGCATGCGCTTTGGGGGCAGA
Phenotype
MIM: 608989
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SDR16C5 PubMed Links

Gene Details

Gene
SDR16C5
Gene Name
short chain dehydrogenase/reductase family 16C, member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318049.1 Intron NP_001304978.1
NM_001318050.1 Intron NP_001304979.1
NM_138969.3 Intron NP_620419.2
XM_011517479.1 Intron XP_011515781.1

View Full Product Details