Product Details
- SNP ID
-
rs640016
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:78102895 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAACCTGGGAGGCGGAGGTTTCAGT[T/C]AGCTGAGATCGTGCCATTGCACTCC
- Phenotype
-
MIM: 608103
- Polymorphism
- T/C, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ALG8
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs60389376] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ALG8
- Gene Name
- ALG8, alpha-1,3-glucosyltransferase
View Full Product Details