Product Details

SNP ID
rs4626463
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:105151796 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAACTAGGGACTTTCCACACTAACC[A/G]CTGAAGTATATTTTCTTTGTCTTGC
Phenotype
MIM: 605824
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BVES-AS1 PubMed Links

Gene Details

Gene
BVES-AS1
Gene Name
BVES antisense RNA 1
There are no transcripts associated with this gene.

Gene
POPDC3
Gene Name
popeye domain containing 3
There are no transcripts associated with this gene.

View Full Product Details