Product Details

SNP ID
rs3092964
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:46354094 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAACTTTATTTCCTACTTCTGCAT[A/G]CCAAGTTTCTACCTCTAGATCTGTT
Phenotype
MIM: 601267
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCR2 PubMed Links

Gene Details

Gene
CCR2
Gene Name
C-C motif chemokine receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001123041.2 351 UTR 5 NP_001116513.2
NM_001123396.1 351 UTR 5 NP_001116868.1
XM_011534069.1 351 Intron XP_011532371.1

View Full Product Details