Product Details

SNP ID
rs138841240
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:73123083 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATACCCTTAGATGTGTAGTAATTT[A/G]ATTTCCTGTTTTGAATATCTTGTAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC105379030 PubMed Links

Gene Details

Gene
LOC105379030
Gene Name
uncharacterized LOC105379030
There are no transcripts associated with this gene.

Gene
TMEM171
Gene Name
transmembrane protein 171
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001161342.2 Intron NP_001154814.1
NM_173490.7 Intron NP_775761.4
XM_011543156.1 Intron XP_011541458.1

View Full Product Details