Product Details

SNP ID
rs4789537
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:78114644 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACAGGAGTTCTGTCAAGAGCCACA[C/T]GGATTGGCTCTGGGCACCTGGCAAC
Phenotype
MIM: 605828 MIM: 610741
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMC6 PubMed Links

Gene Details

Gene
TMC6
Gene Name
transmembrane channel like 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127198.2 Intron NP_001120670.1
NM_001321185.1 Intron NP_001308114.1
NM_007267.7 Intron NP_009198.4
Gene
TNRC6C
Gene Name
trinucleotide repeat containing 6C
There are no transcripts associated with this gene.

Gene
TNRC6C-AS1
Gene Name
TNRC6C antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details