Product Details

SNP ID
rs4747034
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:70479379 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGAGCATGGCCAGGAGCTAGGGTT[C/T]CCCGCGAAGTTAGGGGGAGATTGGG
Phenotype
MIM: 614656
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PALD1 PubMed Links

Gene Details

Gene
PALD1
Gene Name
phosphatase domain containing, paladin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014431.2 Intron NP_055246.2
XM_011539637.2 Intron XP_011537939.1
XM_011539638.2 Intron XP_011537940.1
XM_017016072.1 Intron XP_016871561.1

View Full Product Details