Product Details

SNP ID
rs6572610
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:49772195 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATAGTAATATTTAAAATAGATGTAG[A/G]GGCTTCAGATCCAGAATGGAAAAAT
Phenotype
MIM: 611280 MIM: 608378
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KLHDC2 PubMed Links

Gene Details

Gene
KLHDC2
Gene Name
kelch domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014315.2 Intron NP_055130.1
XM_006720094.3 Intron XP_006720157.1
XM_011536610.1 Intron XP_011534912.1
Gene
NEMF
Gene Name
nuclear export mediator factor
There are no transcripts associated with this gene.

View Full Product Details