Product Details

SNP ID
rs7192854
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:29306522 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCCTGAAAAGTGACCCTGGAGCTG[C/T]GCTTCCCAGGATGTTCAAGGAGTGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SNX29P2 PubMed Links

Gene Details

Gene
SNX29P2
Gene Name
sorting nexin 29 pseudogene 2
There are no transcripts associated with this gene.

View Full Product Details