Product Details
- SNP ID
-
rs7567378
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:68368807 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCTAGAGGTTTATGGAGCAAGCTGT[C/T]GTTTCCAGGACAACCAACCTCACAA
- Phenotype
-
MIM: 173570
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LOC101927723
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75730120] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LOC101927723
- Gene Name
- uncharacterized LOC101927723
There are no transcripts associated with this gene.
- Gene
- PLEK
- Gene Name
- pleckstrin
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002664.2 |
|
Intron |
|
|
NP_002655.2 |
View Full Product Details