Product Details
- SNP ID
-
rs7075733
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:127738311 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCGGCCTTTCCCAGGGCTCCCTTCC[G/T]CCTCCTTTCTGGATTGTCACTTTCT
- Phenotype
-
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
FOXI2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs150430747] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FOXI2
- Gene Name
- forkhead box I2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_207426.2 |
|
Intron |
|
|
NP_997309.2 |
- Gene
- LOC387720
- Gene Name
- collagen alpha-1(X) chain
There are no transcripts associated with this gene.
View Full Product Details