Product Details

SNP ID
rs6553483
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:169987294 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTGATGCTCAAAATATGTGAAAAT[A/G]CAATGAGATTTGGTAGATATTTTAA
Phenotype
MIM: 616523
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MFAP3L PubMed Links

Gene Details

Gene
MFAP3L
Gene Name
microfibrillar associated protein 3 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009554.3 5522 UTR 3 NP_001009554.1
NM_001301647.1 5522 UTR 3 NP_001288576.1
NM_001301648.1 5522 UTR 3 NP_001288577.1
NM_021647.7 5522 UTR 3 NP_067679.6
XM_005263366.3 5522 UTR 3 XP_005263423.1
XM_005263368.3 5522 UTR 3 XP_005263425.1
XM_017008864.1 5522 UTR 3 XP_016864353.1
XM_017008865.1 5522 UTR 3 XP_016864354.1
XM_017008866.1 5522 UTR 3 XP_016864355.1
XM_017008867.1 5522 UTR 3 XP_016864356.1
XM_017008868.1 5522 UTR 3 XP_016864357.1
XM_017008869.1 5522 UTR 3 XP_016864358.1
XM_017008870.1 5522 UTR 3 XP_016864359.1
XM_017008871.1 5522 UTR 3 XP_016864360.1

View Full Product Details