Product Details

SNP ID
rs6919872
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:138405046 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGGGGCGGTGCAGCCCCTAGACAGG[A/C]CGGCTCCAGGTCGACCCGAGATCAT
Phenotype
MIM: 605825
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HEBP2 PubMed Links

Gene Details

Gene
HEBP2
Gene Name
heme binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001326380.1 Intron NP_001313309.1
NM_001326381.1 Intron NP_001313310.1
NM_014320.2 Intron NP_055135.1
XM_017010631.1 Intron XP_016866120.1

View Full Product Details