Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001002862.2 | Intron | NP_001002862.1 | |||
NM_001135751.1 | Intron | NP_001129223.1 | |||
NM_198440.3 | Intron | NP_940842.2 | |||
XM_011530505.2 | Intron | XP_011528807.1 | |||
XM_011530506.2 | Intron | XP_011528808.1 | |||
XM_017029078.1 | Intron | XP_016884567.1 | |||
XM_017029079.1 | Intron | XP_016884568.1 | |||
XM_017029080.1 | Intron | XP_016884569.1 | |||
XM_017029081.1 | Intron | XP_016884570.1 | |||
XM_017029082.1 | Intron | XP_016884571.1 |