Product Details

SNP ID
rs9857750
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:159072746 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAGTCCTCATTGACTATTAATATC[C/G]ACATTCTACTGGTGAAGAAACTGAG
Phenotype
MIM: 611622
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
IQCJ PubMed Links

Gene Details

Gene
IQCJ
Gene Name
IQ motif containing J
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042705.2 Intron NP_001036170.1
NM_001042706.2 Intron NP_001036171.1
NM_001197100.1 Intron NP_001184029.1
Gene
IQCJ-SCHIP1
Gene Name
IQCJ-SCHIP1 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001197113.1 Intron NP_001184042.1
NM_001197114.1 Intron NP_001184043.1

View Full Product Details