Product Details

SNP ID
rs7545064
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:119572448 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTTCTTCACGCAGTCCAATGTAAA[A/G]AGCAATAAATGTTTTAATCCTTAAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HSD3BP4 PubMed Links

Gene Details

Gene
HSD3BP4
Gene Name
hydroxy-delta-5-steroid dehydrogenase, 3 beta, pseudogene 4
There are no transcripts associated with this gene.

View Full Product Details