Product Details

SNP ID
rs6116933
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:5751065 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTTGACAGAATTGTGCATCTATT[A/G]CTGTGATCTCATTTTAGAATACTTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C20orf196 PubMed Links

Gene Details

Gene
C20orf196
Gene Name
chromosome 20 open reading frame 196
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303477.1 Intron NP_001290406.1
NM_001303478.1 Intron NP_001290407.1
NM_001303479.1 Intron NP_001290408.1
NM_152504.3 Intron NP_689717.2
XM_006723549.3 Intron XP_006723612.1
XM_011529177.2 Intron XP_011527479.1
XM_011529178.2 Intron XP_011527480.1
XM_011529179.2 Intron XP_011527481.1
XM_011529180.2 Intron XP_011527482.1
XM_011529181.2 Intron XP_011527483.1
XM_011529182.2 Intron XP_011527484.1
XM_017027684.1 Intron XP_016883173.1

View Full Product Details