Product Details
- SNP ID
-
rs9901901
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:28283487 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGTTAAGGCATATTGGTTAAAAGCA[C/T]AGGTTTCAGTACCATGATTCTACCA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KRT18P55
PubMed Links
Gene Details
- Gene
- KRT18P55
- Gene Name
- keratin 18 pseudogene 55
There are no transcripts associated with this gene.
View Full Product Details