Product Details
- SNP ID
-
rs6572612
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:49775604 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AAGGAGTAGCCAAGATACAGAGATA[C/T]AAGCAAATGTCTTTTTTTTTTTTTT
- Phenotype
-
MIM: 611280
MIM: 608378
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
KLHDC2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs8007202] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- KLHDC2
- Gene Name
- kelch domain containing 2
- Gene
- NEMF
- Gene Name
- nuclear export mediator factor
There are no transcripts associated with this gene.
View Full Product Details