Product Details

SNP ID
rs9485189
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:147509873 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGAGTGGAAAGTCAGGCCCTGCCT[C/T]TCTCAATTCCATAAAAAATCCCAGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SAMD5 PubMed Links

Gene Details

Gene
SAMD5
Gene Name
sterile alpha motif domain containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001030060.2 Intron NP_001025231.1
XM_017010850.1 Intron XP_016866339.1

View Full Product Details