Product Details

SNP ID
rs5963386
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:38150099 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAACTTGCTAGAAATGGAGACTCT[C/T]ATGCCCAACCCCAAACCTACTGAAT
Phenotype
MIM: 300187
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SRPX PubMed Links

Gene Details

Gene
SRPX
Gene Name
sushi repeat containing protein, X-linked
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170750.1 Intron NP_001164221.1
NM_001170751.1 Intron NP_001164222.1
NM_001170752.1 Intron NP_001164223.1
NM_006307.4 Intron NP_006298.1
XM_017029893.1 Intron XP_016885382.1

View Full Product Details