Product Details

SNP ID
rs7317207
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:41219619 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAAAGGTGGCATCAGGGAAAGATTC[A/G]GGAAGCCAGAAAGTTTTGAATTTGG
Phenotype
MIM: 604601
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC101929140 PubMed Links

Gene Details

Gene
LOC101929140
Gene Name
uncharacterized LOC101929140
There are no transcripts associated with this gene.

Gene
MTRF1
Gene Name
mitochondrial translational release factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004294.2 Intron NP_004285.2
XM_005266599.2 Intron XP_005266656.1
XM_006719897.2 Intron XP_006719960.1
XM_006719898.2 Intron XP_006719961.1
XM_006719900.2 Intron XP_006719963.1
XM_011535315.2 Intron XP_011533617.1
XM_011535317.2 Intron XP_011533619.1
XM_011535318.2 Intron XP_011533620.1
XM_011535320.2 Intron XP_011533622.1
XM_011535321.2 Intron XP_011533623.1
XM_011535323.2 Intron XP_011533625.1
XM_011535325.2 Intron XP_011533627.1
XM_011535326.2 Intron XP_011533628.1
XM_011535327.2 Intron XP_011533629.1
XM_017020856.1 Intron XP_016876345.1
XM_017020857.1 Intron XP_016876346.1
XM_017020858.1 Intron XP_016876347.1
XM_017020859.1 Intron XP_016876348.1
XM_017020860.1 Intron XP_016876349.1
XM_017020861.1 Intron XP_016876350.1
XM_017020862.1 Intron XP_016876351.1
XM_017020863.1 Intron XP_016876352.1
XM_017020864.1 Intron XP_016876353.1
XM_017020865.1 Intron XP_016876354.1
XM_017020866.1 Intron XP_016876355.1
XM_017020867.1 Intron XP_016876356.1
XM_017020868.1 Intron XP_016876357.1
XM_017020869.1 Intron XP_016876358.1
XM_017020870.1 Intron XP_016876359.1
XM_017020871.1 Intron XP_016876360.1
XM_017020872.1 Intron XP_016876361.1
XM_017020873.1 Intron XP_016876362.1
XM_017020874.1 Intron XP_016876363.1
XM_017020875.1 Intron XP_016876364.1
XM_017020876.1 Intron XP_016876365.1
XM_017020877.1 Intron XP_016876366.1
XM_017020878.1 Intron XP_016876367.1
XM_017020879.1 Intron XP_016876368.1

View Full Product Details