Product Details

SNP ID
rs6891719
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:95856218 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTACCGTCAGCTCCTACCATTGTA[C/T]ACTCCTGCCTGCAGCTTTTTTTCTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC01554 PubMed Links

Gene Details

Gene
LINC01554
Gene Name
long intergenic non-protein coding RNA 1554
There are no transcripts associated with this gene.

Gene
LOC102724720
Gene Name
uncharacterized LOC102724720
There are no transcripts associated with this gene.

View Full Product Details