Product Details

SNP ID
rs10886403
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:119144871 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGTAGCACACGTAGCTCATGGCTAC[C/G]ATAATTGGACAGCACCAATCTATTA
Phenotype
MIM: 615564
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FAM45A PubMed Links

Gene Details

Gene
FAM45A
Gene Name
family with sequence similarity 45 member A
There are no transcripts associated with this gene.

Gene
SFXN4
Gene Name
sideroflexin 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_213649.1 Intron NP_998814.1
XM_005269525.4 Intron XP_005269582.1
XM_005269526.2 Intron XP_005269583.1
XM_005269527.1 Intron XP_005269584.1
XM_011539282.2 Intron XP_011537584.1

View Full Product Details