Product Details

SNP ID
rs12386864
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:67428936 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGAATGTTGAATAAGCATAAATC[G/T]GAAAGAGTATTAAGATAATTACATT
Phenotype
MIM: 609562
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CPA6 PubMed Links

Gene Details

Gene
CPA6
Gene Name
carboxypeptidase A6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020361.4 Intron NP_065094.3
XM_011517570.2 Intron XP_011515872.1
XM_017013646.1 Intron XP_016869135.1
XM_017013647.1 Intron XP_016869136.1
Gene
LOC102724708
Gene Name
uncharacterized LOC102724708
There are no transcripts associated with this gene.

View Full Product Details