Product Details

SNP ID
rs7166434
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:45512915 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGCCGAGGTGGGAGAACTGCTTGAG[C/T]GTAAGAGTCTGAGGCTGCAGTAAGC
Phenotype
MIM: 611314 MIM: 602095
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HMGN2P46 PubMed Links

Gene Details

Gene
HMGN2P46
Gene Name
high mobility group nucleosomal binding domain 2 pseudogene 46
There are no transcripts associated with this gene.

Gene
SLC30A4
Gene Name
solute carrier family 30 member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321036.1 Intron NP_001307965.1
NM_013309.5 Intron NP_037441.2
XM_011521997.2 Intron XP_011520299.1
XM_017022560.1 Intron XP_016878049.1
XM_017022561.1 Intron XP_016878050.1
XM_017022562.1 Intron XP_016878051.1

View Full Product Details