Product Details

SNP ID
rs12119002
Assay Type
Functionally tested
NCBI dbSNP Submissions
29
Location
Chr.1:229274748 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATGTTCAGAAAGGAGCTGAAAAGT[A/G]TATGTAGTGGGAAAATGCCAGGAAT
Phenotype
MIM: 179511
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RAB4A PubMed Links

Gene Details

Gene
RAB4A
Gene Name
RAB4A, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271998.1 Intron NP_001258927.1
NM_004578.3 Intron NP_004569.2

View Full Product Details