Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001206794.1 | Intron | NP_001193723.1 | |||
NM_016374.5 | Intron | NP_057458.4 | |||
NM_031371.3 | Intron | NP_112739.2 | |||
XM_006711781.2 | Intron | XP_006711844.1 | |||
XM_011544212.2 | Intron | XP_011542514.1 | |||
XM_017001468.1 | Intron | XP_016856957.1 | |||
XM_017001469.1 | Intron | XP_016856958.1 | |||
XM_017001470.1 | Intron | XP_016856959.1 | |||
XM_017001471.1 | Intron | XP_016856960.1 | |||
XM_017001472.1 | Intron | XP_016856961.1 |