Product Details

SNP ID
rs11227352
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:65917141 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCAGGAGGAAGGGGAGGTATAGCG[A/G]GGGATGAGCGTTCCAAGAAGTCTCT
Phenotype
MIM: 602289
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C11orf68 PubMed Links

Gene Details

Gene
C11orf68
Gene Name
chromosome 11 open reading frame 68
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135635.1 1226 UTR 3 NP_001129107.1
NM_031450.3 1226 UTR 3 NP_113638.2
Gene
DRAP1
Gene Name
DR1 associated protein 1
There are no transcripts associated with this gene.

View Full Product Details