Product Details

SNP ID
rs12791960
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:12377682 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCCGCAGAAGTCGCCTTCTGTCC[A/C]CAAGTCTCCCACTCCCAAGTCGCCC
Phenotype
MIM: 608120
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PARVA PubMed Links

Gene Details

Gene
PARVA
Gene Name
parvin alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018222.4 204 Missense Mutation CAC,CCC H52P NP_060692.2
XM_005253015.3 204 Intron XP_005253072.1

View Full Product Details