Product Details

SNP ID
rs28936671
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:36517757 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGCCTGGCGCTTCATTTTGTAGC[T/G]GTGGTTCTGGAACCAGATCTTGACC
Phenotype
MIM: 600635
Polymorphism
T/G, Transversion substitution
Allele Nomenclature
Literature Links
NKX2-1 PubMed Links

Gene Details

Gene
NKX2-1
Gene Name
NK2 homeobox 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001079668.2 978 Missense Mutation AGC,CGC S243R NP_001073136.1
NM_003317.3 978 Missense Mutation AGC,CGC S213R NP_003308.1
Gene
NKX2-1-AS1
Gene Name
NKX2-1 antisense RNA 1
There are no transcripts associated with this gene.

Gene
SFTA3
Gene Name
surfactant associated 3
There are no transcripts associated with this gene.

View Full Product Details