Product Details

SNP ID
rs16987299
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:56515682 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGAAGTAAATCTGAAGGTGAAAAAT[A/G]CCCACAATAACTTTTAAAATTTCTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF471 PubMed Links

Gene Details

Gene
ZNF471
Gene Name
zinc finger protein 471
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321768.1 Intron NP_001308697.1
NM_020813.3 Intron NP_065864.2
XM_011527148.1 Intron XP_011525450.1
XM_017027032.1 Intron XP_016882521.1

View Full Product Details