Product Details
- SNP ID
-
rs5986186
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:52653044 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTCAGCTAGACATGTCTGCCATTCA[G/T]CTGGAGCCCCTTCCTGTGTGCTGGA
- Phenotype
-
MIM: 300542
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
SSX7
PubMed Links
Gene Details
- Gene
- SSX7
- Gene Name
- SSX family member 7
View Full Product Details