Product Details

SNP ID
rs16979502
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:37377650 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGATCATGCTGTACTCTGGGGCTCA[C/T]AACAGCATAGCTCTCTTACTAACCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF527 PubMed Links

Gene Details

Gene
ZNF527
Gene Name
zinc finger protein 527
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032453.1 Intron NP_115829.1
XM_005259328.4 Intron XP_005259385.1
XM_005259329.3 Intron XP_005259386.1
XM_017027380.1 Intron XP_016882869.1
XM_017027381.1 Intron XP_016882870.1

View Full Product Details