Product Details
- SNP ID
-
rs7913553
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:68733532 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTCTCTTTCTTTAAATTTTAAAAAT[C/T]TGGTTTTGGCCTGAAATCCCAGCAC
- Phenotype
-
MIM: 612569
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CCAR1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs112455345] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CCAR1
- Gene Name
- cell division cycle and apoptosis regulator 1
View Full Product Details