Product Details
- SNP ID
-
rs6501819
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:75517353 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCCGGTCTTTAGAGAACTGATAACC[A/G]AACTTCTAAAGCAGGAGGTGGTACG
- Phenotype
-
MIM: 612185
MIM: 608755
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CASKIN2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs11316466] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CASKIN2
- Gene Name
- CASK interacting protein 2
There are no transcripts associated with this gene.
- Gene
- LLGL2
- Gene Name
- LLGL2, scribble cell polarity complex component
There are no transcripts associated with this gene.
- Gene
- TSEN54
- Gene Name
- tRNA splicing endonuclease subunit 54
View Full Product Details