Product Details

SNP ID
rs9964567
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:35301756 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGAATTCTGTTTAGTATTTATTTT[A/G]GTGCAGGTATATTTTTAAAAACCTT
Phenotype
MIM: 604754
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF271P PubMed Links

Gene Details

Gene
ZNF271P
Gene Name
zinc finger protein 271, pseudogene
There are no transcripts associated with this gene.

View Full Product Details