Product Details
- SNP ID
-
rs4524063
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:23236226 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCCAGTGTCCTTTCTTGACTCCTA[C/T]CACAGGGCTCACAGCTGGCAGCTCA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF724P
PubMed Links
Gene Details
- Gene
- ZNF724P
- Gene Name
- zinc finger protein 724, pseudogene
There are no transcripts associated with this gene.
View Full Product Details