Product Details

SNP ID
rs11542705
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:26863677 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCGGTCAGCCTACCAGGAGGCCAT[A/G]GACATCAGCAAGAAGGAGATGCCGC
Phenotype
MIM: 601290
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SFN PubMed Links

Gene Details

Gene
SFN
Gene Name
stratifin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006142.3 536 Missense Mutation ATA,ATG I155M NP_006133.1
Gene
ZDHHC18
Gene Name
zinc finger DHHC-type containing 18
There are no transcripts associated with this gene.

View Full Product Details