Product Details

SNP ID
hCV60080750
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:50560527 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAATTAAAATTAGTACCAAGGAAA[A/C]AAAAGAAAAAGAAAAAAAAAGAAAA
Phenotype
MIM: 605692
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TRPM7 PubMed Links

Gene Details

Gene
TRPM7
Gene Name
transient receptor potential cation channel subfamily M member 7
There are no transcripts associated with this gene.

View Full Product Details