Product Details
- SNP ID
-
rs28676928
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:20515222 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGCATGCTATGCCAAGCCCTGTTGC[A/T]GGAGTTGACAGATGTTGACTCATCC
- Phenotype
-
MIM: 607372
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
MED15
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116522857] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MED15
- Gene Name
- mediator complex subunit 15
View Full Product Details