Product Details

SNP ID
rs17879610
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:134471087 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTGATTTTTTGTTTGTATATTTT[C/G]CTTTAATTTAATGTATCCTTTAAAA
Phenotype
MIM: 308000
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HPRT1 PubMed Links

Gene Details

Gene
HPRT1
Gene Name
hypoxanthine phosphoribosyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000194.2 Intron NP_000185.1

View Full Product Details