Product Details
- SNP ID
-
rs72884533
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:23463142 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAGCAGAGTTTTAGGATTTGATGAG[C/T]AATTGTCATGACAAGTGGTTTGGAA
- Phenotype
-
MIM: 603579
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
RIOK3
PubMed Links
Gene Details
- Gene
- RIOK3
- Gene Name
- RIO kinase 3
View Full Product Details