Product Details

SNP ID
rs45496096
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:35101148 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCTCGAAGACATCTGTGGGTATG[A/G]AAACCACCCTCCAGGGCCCAAGATT
Phenotype
MIM: 602954
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RAD51D PubMed Links

Gene Details

Gene
RAD51D
Gene Name
RAD51 paralog D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142571.1 Intron NP_001136043.1
NM_002878.3 Intron NP_002869.3
NM_133629.2 Intron NP_598332.1
Gene
RAD51L3-RFFL
Gene Name
RAD51L3-RFFL readthrough
There are no transcripts associated with this gene.

View Full Product Details