Product Details

SNP ID
rs41294792
Assay Type
Functionally tested
NCBI dbSNP Submissions
17
Location
Chr.1:54139569 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAATAGTGGAAACAAGCGGGAGCC[A/G]TACCGTCCAGTCTTAGGGGTCCCGA
Phenotype
MIM: 612320
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDCP2 PubMed Links

Gene Details

Gene
CDCP2
Gene Name
CUB domain containing protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_201546.3 1385 Missense Mutation ACG,ATG T434M NP_963840.2

View Full Product Details