Product Details

SNP ID
rs62278494
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:142726846 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTAAACATTCACTAAAGCAGCTA[C/T]TGGGGTCGAGTGTTCATGCTTTTCA
Phenotype
MIM: 602343
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TRPC1 PubMed Links

Gene Details

Gene
TRPC1
Gene Name
transient receptor potential cation channel subfamily C member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001251845.1 Intron NP_001238774.1
NM_003304.4 Intron NP_003295.1
XM_005247738.3 Intron XP_005247795.1
XM_005247739.2 Intron XP_005247796.1
XM_017007121.1 Intron XP_016862610.1
XM_017007122.1 Intron XP_016862611.1

View Full Product Details